3-Fluorbenzhydrazid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Fluorbenzoesäurehydrazid, 3-Fluorbenzol-1-carbohydrazid, 3-Fluorbenzohydrazid
PC3495F-5G
PC3495F-25G
APOSPC3495F-5GEA
58
EUR
APOSPC3495F-5G
APOSPC3495F-25G
3-Fluorbenzhydrazid 98%
3-Fluorbenzhydrazid
Formel:
C₇H₇FN₂O Molecular Weight: 154,14 g/mol Schmelzpunkt: 138…139 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00060561 CAS-Nummer: 499-55-8 EINECS: 610-486-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...