4-Fluorbenzhydrazid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Fluorbenzol-1-carbohydrazid, 4-Fluorbenzohydrazid, 4-Fluorbenzoesäurehydrazid

PC3495G-100G PC3495G-25G PC3495G-5G
APOSPC3495G-100GEA 430 EUR
APOSPC3495G-100G APOSPC3495G-25G APOSPC3495G-5G
4-Fluorbenzhydrazid 98%
4-Fluorbenzhydrazid
Formel: C₇H₇FN₂O
Molecular Weight: 154,14 g/mol
Schmelzpunkt: 160…166 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00060562
CAS-Nummer: 456-06-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR