1-[4-Fluoro-2-(methylsulphonyl)phenyl]hydrazine 95+%

Lieferant: Apollo Scientific

PC3505-1G PC3505-5G
APOSPC3505-1GEA 592 EUR
APOSPC3505-1G APOSPC3505-5G
1-[4-Fluoro-2-(methylsulphonyl)phenyl]hydrazine 95+%
1-[4-Fluoro-2-(methylsulphonyl)phenyl]hydrazine
Formel: C₇H₉FN₂O₂S
Molecular Weight: 204.22 g/mol
Schmelzpunkt: 117…120 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08692478
CAS-Nummer: 914637-59-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR