2-(2,2,2-Trifluoroethoxy)pyridine-3-carbonitrile 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(2,2,2-Trifluoroethoxy)nicotinonitrile
PC3506-1G
PC3506-5G
APOSPC3506-1GEA
109
EUR
APOSPC3506-1G
APOSPC3506-5G
2-(2,2,2-Trifluoroethoxy)pyridine-3-carbonitrile 95%
2-(2,2,2-Trifluoroethoxy)pyridine-3-carbonitrile
Formel:
C₈H₅F₃N₂O Molecular Weight: 202,14 g/mol Schmelzpunkt: 44…46 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00275529 CAS-Nummer: 175277-89-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...