3-Fluor-4-(methylcarbamoyl)phenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Fluoro-4-(methylcarbamoyl)benzeneboronic acid, (3-Fluoro-4-(methylcarbamoyl)phenyl)boronic acid
PC3532-1G
PC3532-5G
APOSPC3532-1GEA
215
EUR
APOSPC3532-1G
APOSPC3532-5G
3-Fluor-4-(methylcarbamoyl)phenylboronsäure 98%
3-Fluor-4-(methylcarbamoyl)phenylboronsäure
Formel:
C₈H₉BFNO₃ Schmelzpunkt: 173…175 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08436026 CAS-Nummer: 849833-86-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...