5-Carboxy-2-fluorphenylboronsäure 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Borono-4-fluorobenzoic acid
PC3563-1G
PC3563-5G
APOSPC3563-1GEA
75.4
EUR
APOSPC3563-1G
APOSPC3563-5G
5-Carboxy-2-fluorphenylboronsäure 96%
5-Carboxy-2-fluorphenylboronsäure
Formel:
C₇H₆BFO₄ Molecular Weight: 183,9300 g/mol Schmelzpunkt: 224…226 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08436065 CAS-Nummer: 874219-59-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...