4-(Dimethylcarbamoyl)-3-fluorphenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Dimethylcarbamoyl-3-fluorobenzeneboronic acid
PC3564-1G
PC3564-5G
APOSPC3564-1GEA
220
EUR
APOSPC3564-1G
APOSPC3564-5G
4-(Dimethylcarbamoyl)-3-fluorphenylboronsäure 98%
4-(Dimethylcarbamoyl)-3-fluorphenylboronsäure
Formel:
C₉H₁₁BFNO₃ Molecular Weight: 211.00 g/mol Schmelzpunkt: 147…149 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08436060 CAS-Nummer: 874289-13-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...