3-Fluor-4-(methoxycarbamoyl)phenylboronsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3-Fluoro-4-(methoxycarbamoyl)phenyl)boronic acid, 3-Fluoro-4-(methoxycarbamoyl)benzeneboronic acid
PC3565-1G
PC3565-5G
APOSPC3565-1GEA
258
EUR
APOSPC3565-1G
APOSPC3565-5G
3-Fluor-4-(methoxycarbamoyl)phenylboronsäure 97%
3-Fluor-4-(methoxycarbamoyl)phenylboronsäure
Formel:
C₈H₉BFNO₄ Molecular Weight: 212,97 g/mol Schmelzpunkt: 161…163 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08436059 CAS-Nummer: 913835-58-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...