3-Brom-5-(trifluormethyl)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Bromo-5-(trifluoromethyl)benzeneboronic acid

PC3571-1G PC3571-5G
APOSPC3571-1GEA 94.4 EUR
APOSPC3571-1G APOSPC3571-5G
3-Brom-5-(trifluormethyl)phenylboronsäure 98%
3-Brom-5-(trifluormethyl)phenylboronsäure
Formel: C₇H₅BBrF₃O₂
Molecular Weight: 268.82 g/mol
Schmelzpunkt: 177…180 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08436051
CAS-Nummer: 913835-64-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR