5-(tert-Butylcarbamoyl)-2-fluorphenylboronsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-(Tert-butylcarbmoyl)-2-fluorophenylboronic acid, 5-tert-Butylcarbamoyl-2-fluorobenzeneboronic acid, (5-(tert-Butylcarbamoyl)-2-fluorophenyl)boronic acid
PC3576-1G
PC3576-5G
APOSPC3576-1GEA
277
EUR
APOSPC3576-1G
APOSPC3576-5G
5-(tert-Butylcarbamoyl)-2-fluorphenylboronsäure 97%
5-(tert-Butylcarbamoyl)-2-fluorphenylboronsäure
Formel:
C₁₁H₁₅BFNO₃ Molecular Weight: 239,05 g/mol Schmelzpunkt: 142…144 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08436054 CAS-Nummer: 874289-51-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...