3-Fluor-4-(piperidin-1-carbonyl)phenylboronsäure 98%

Lieferant: Apollo Scientific

PC3577-10G PC3577-1G PC3577-5G
APOSPC3577-10GEA 734 EUR
APOSPC3577-10G APOSPC3577-1G APOSPC3577-5G
3-Fluor-4-(piperidin-1-carbonyl)phenylboronsäure 98%
3-Fluor-4-(piperidin-1-carbonyl)phenylboronsäure
Formel: C₁₂H₁₅BFNO₃
Schmelzpunkt: 162…164 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08436039
CAS-Nummer: 874289-10-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR