4-(Cyclohexylcarbamoyl)-3-fluorphenylboronsäure 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-(Cyclohexylcarbamoyl)-3-fluorophenyl)boronic acid
PC3579-1G
PC3579-5G
APOSPC3579-1GEA
136
EUR
APOSPC3579-1G
APOSPC3579-5G
4-(Cyclohexylcarbamoyl)-3-fluorphenylboronsäure 95%
4-(Cyclohexylcarbamoyl)-3-fluorphenylboronsäure
Formel:
C₁₃H₁₇BFNO₃ Molecular Weight: 265.09 g/mol Schmelzpunkt: 157…159 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08436036 CAS-Nummer: 874289-11-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...