2-Fluor-5-(1-pyrrolidinylcarbonyl)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-Fluoro-5-(pyrrolidine-1-carbonyl)phenyl)boronic acid, 2-Fluoro-5-(pyrolidine-1-carbonyl)phenylboronic acid

PC3581-1G PC3581-5G
APOSPC3581-1GEA 241 EUR
APOSPC3581-1G APOSPC3581-5G
2-Fluor-5-(1-pyrrolidinylcarbonyl)phenylboronsäure 98%
2-Fluor-5-(1-pyrrolidinylcarbonyl)phenylboronsäure
Formel: C₁₁H₁₃BFNO₃
Schmelzpunkt: 146…148 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08436030
CAS-Nummer: 874289-42-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR