2-Fluor-5-(methoxy(methyl)carbamoyl)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-Methoxy-N-methyl 3-borono-4-fluorobenzamide

PC3588-1G PC3588-5G
APOSPC3588-1GEA 380 EUR
APOSPC3588-1G APOSPC3588-5G
2-Fluor-5-(methoxy(methyl)carbamoyl)phenylboronsäure 98%
2-Fluor-5-(methoxy(methyl)carbamoyl)phenylboronsäure
Formel: C₉H₁₁BFNO₄
Molecular Weight: 227,0000 g/mol
Schmelzpunkt: 117…119 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08436038
CAS-Nummer: 874289-59-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR