3-Fluor-2-methoxy-5-pyridinboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(5-Fluoro-6-methoxypyridin-3-yl)boronic acid, 3-Fluoro-2-methoxypyridine-5-boronic acid
PC3612-1G
APOSPC3612-1GEA
75.4
EUR
APOSPC3612-1G
3-Fluor-2-methoxy-5-pyridinboronsäure
3-Fluor-2-methoxy-5-pyridinboronsäure
Formel:
C₆H₇BFNO₃ Molecular Weight: 170,9341 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD04112520 CAS-Nummer: 856250-60-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...