5-Bromo-2-fluorobenzeneboronic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-2-fluorophenylboronic acid, 5-Bromo-2-fluorobenzeneboronic acid
PC3621-100G
PC3621-25G
APOSPC3621-100GEA
434
EUR
APOSPC3621-100G
APOSPC3621-25G
5-Bromo-2-fluorobenzeneboronic acid 98%
5-Bromo-2-fluorobenzeneboronic acid
Formel:
C₆H₅BBrFO₂ Molecular Weight: 218,82 g/mol Schmelzpunkt: 220…224 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD05664307 CAS-Nummer: 112204-57-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...