4-(tert-Butylcarbamoyl)-3-fluorphenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-(tert-Butylcarbamoyl)-3-fluorophenyl)boronic acid

PC3623-1G PC3623-5G
APOSPC3623-1GEA 241 EUR
APOSPC3623-1G APOSPC3623-5G
4-(tert-Butylcarbamoyl)-3-fluorphenylboronsäure 98%
4-(tert-Butylcarbamoyl)-3-fluorphenylboronsäure
Formel: C₁₁H₁₅BFNO₃
Molecular Weight: 239,0500 g/mol
Schmelzpunkt: 140…143 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08689525
CAS-Nummer: 874289-18-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR