7-Bromo-5-fluorobenzo[b]furan 97%

Lieferant: Apollo Scientific
Achtung

PC3665-1G
APOSPC3665-1GEA 112 EUR
APOSPC3665-1G
7-Bromo-5-fluorobenzo[b]furan 97%
7-Bromo-5-fluorobenzo[b]furan
Formel: C₈H₄BrFO
Molecular Weight: 215,021 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09056780
CAS-Nummer: 253429-19-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR