3-{4-[5-(Trifluoromethyl)pyridin-2-yl]piperazin-1-yl}propanoic acid
Lieferant: Apollo Scientific
PC3671-1G
PC3671-10G
PC3671-5G
APOSPC3671-1GEA
125
EUR
APOSPC3671-1G
APOSPC3671-10G
APOSPC3671-5G
3-{4-[5-(Trifluoromethyl)pyridin-2-yl]piperazin-1-yl}propanoic acid
3-{4-[5-(Trifluoromethyl)pyridin-2-yl]piperazin-1-yl}propanoic acid
Formel:
C₁₃H₁₆F₃N₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11035862 CAS-Nummer: 1031929-07-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...