6-Fluor-2-pyridincarbonitril

Lieferant: Apollo Scientific
Gefahr

Synonyme: 6-Fluor-2-pyridincarbonsäurenitril, 2-Cyan-6-fluorpyridin

PC3684-10G PC3684-1G PC3684-5G
APOSPC3684-10GEA 136 EUR
APOSPC3684-10G APOSPC3684-1G APOSPC3684-5G
6-Fluor-2-pyridincarbonitril
6-Fluor-2-pyridincarbonitril
Formel: C₆H₃FN₂
Molecular Weight: 122,1 g/mol
Schmelzpunkt: 31…35 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09263323
CAS-Nummer: 3939-15-9
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR