4-Fluor-3-methoxybenzylalkohol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-Fluoro-3-methoxyphenyl)methanol
PC3692-1G
PC3692-5G
PC3692-25G
APOSPC3692-1GEA
24.8
EUR
APOSPC3692-1G
APOSPC3692-5G
APOSPC3692-25G
4-Fluor-3-methoxybenzylalkohol 97%
4-Fluor-3-methoxybenzylalkohol
Formel:
C₈H₉FO₂ Molecular Weight: 156,16 g/mol Schmelzpunkt: 47…50 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00272133 CAS-Nummer: 128495-45-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...