4-(4-Fluorbenzoyl)pyridin

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-Fluorophenyl)(pyridin-4-yl)methanone

PC3699-1G PC3699-25G PC3699-5G
APOSPC3699-1GEA 58.2 EUR
APOSPC3699-1G APOSPC3699-25G APOSPC3699-5G
4-(4-Fluorbenzoyl)pyridin
4-(4-Fluorbenzoyl)pyridin
Formel: C₁₂H₈FNO
Molecular Weight: 201.2 g/mol
Schmelzpunkt: 85…87 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02683088
CAS-Nummer: 41538-36-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR