4,5,5-Trifluoropent-4-en-1-ol 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC3713-1G
PC3713-5G
PC3713-100MG
APOSPC3713-1GEA
136
EUR
APOSPC3713-1G
APOSPC3713-5G
APOSPC3713-100MG
4,5,5-Trifluoropent-4-en-1-ol 95%
4,5,5-Trifluoropent-4-en-1-ol
Formel:
C₅H₇F₃O Siedepunkt: 48 °C/10mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00156073 CAS-Nummer: 109993-33-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...