4-Fluor-3-methylphenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Methyl-4-fluorophenylboronic acid, 4-Fluoro-3-methylbenzeneboronic acid

PC3716-5G PC3716-25G
APOSPC3716-5GEA 21.9 EUR
APOSPC3716-5G APOSPC3716-25G
4-Fluor-3-methylphenylboronsäure
4-Fluor-3-methylphenylboronsäure
Formel: C₇H₈BFO₂
Molecular Weight: 153,95 g/mol
Schmelzpunkt: 211…217 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01863527
CAS-Nummer: 139911-27-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR