Ethyl-2-methyl-6-(trifluoromethyl)nicotinate 95+%
Lieferant: Apollo Scientific
PC3721-10G
PC3721-5G
PC3721-1G
PC3721-250MG
APOSPC3721-10GEA
0
EUR
APOSPC3721-10G
APOSPC3721-5G
APOSPC3721-1G
APOSPC3721-250MG
Ethyl-2-methyl-6-(trifluoromethyl)nicotinate 95+%
Ethyl-2-methyl-6-(trifluoromethyl)nicotinate
Formel:
C₁₀H₁₀F₃NO₂ Molecular Weight: 233,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10688283 CAS-Nummer: 380355-65-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...