N-(4-Fluorobenzyl)urea 97%

Lieferant: Apollo Scientific
Achtung

PC3723E-5G PC3723E-25G
APOSPC3723E-5GEA 159 EUR
APOSPC3723E-5G APOSPC3723E-25G
N-(4-Fluorobenzyl)urea 97%
N-(4-Fluorobenzyl)urea
Formel: C₈H₉FN₂O
Molecular Weight: 168,17 g/mol
Schmelzpunkt: 172…175 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00085086
CAS-Nummer: 76523-24-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR