N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate)

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-(Chloromethyl)-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane Bis(tetrafluoroborate), F-TEDA-BF4, 1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate)

PC3728K-100G PC3728K-250G PC3728K-25G
APOSPC3728K-100GEA 102 EUR
APOSPC3728K-100G APOSPC3728K-250G APOSPC3728K-25G
N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate)
N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate)

Electrophilic fluorinating agent. Ask for technical literature(SelectfluorR - Registered trademark of Air Products).


Decomposes exothermally at >1000 C.

Formel: C₇H₁₄B₂ClF₉N₂
Molecular Weight: 354.26 g/mol
Schmelzpunkt: 260 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00142607
CAS-Nummer: 140681-55-6
EINECS: 414-380-4
UN: 3088
ADR: 4.2 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR