3-Fluorocinnamic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(3-Fluorophenyl)acrylic acid, m-Fluorocinnamic acid, 3-Fluorocinnamic Acid
PC3731-100G
PC3731-25G
PC3731-5G
APOSPC3731-100GEA
65.4
EUR
APOSPC3731-100G
APOSPC3731-25G
APOSPC3731-5G
3-Fluorocinnamic acid 98%
3-Fluorocinnamic acid
Formel:
C₉H₇FO₂ Molecular Weight: 166.15 g/mol Schmelzpunkt: 166…167 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004383 CAS-Nummer: 458-46-8 EINECS: 207-281-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...