5-Fluorcytosin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Flucytosin, 4-Amino-5-fluor-2-hydroxypyrimidin, 4-Amino-5-fluor-2-pyrimidinol, 5-FC
PC3735-100G
PC3735-25G
PC3735-250G
APOSPC3735-100GEA
34.8
EUR
APOSPC3735-100G
APOSPC3735-25G
APOSPC3735-250G
5-Fluorcytosin 98%
5-Fluorcytosin
Formel:
C₄H₄FN₃O Molecular Weight: 129,09 g/mol Schmelzpunkt: 298…300 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00006035 CAS-Nummer: 2022-85-7 EINECS: 217-968-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...