2-Fluor-4-nitroanilin
Lieferant: Apollo Scientific
|
Achtung
|
PC3744-100G
PC3744-25G
PC3744-5G
APOSPC3744-100GEA
62.8
EUR
APOSPC3744-100G
APOSPC3744-25G
APOSPC3744-5G
2-Fluor-4-nitroanilin
2-Fluor-4-nitroanilin
Formel:
C₆H₅FN₂O₂ Molecular Weight: 156.12 g/mol Schmelzpunkt: 122…130 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00034560 CAS-Nummer: 369-35-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...