1-Fluor-2,6-dichlorpyridiniumtetrafluoroborat 96%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2,6-Dichloro-1-fluoropyridin-1-ium tetrafluoroborate, 2,6-Dichloro-N-fluoropyridinium tetrafluoroborate

PC3744E-25G PC3744E-5G
APOSPC3744E-25GEA 0 EUR
APOSPC3744E-25G APOSPC3744E-5G
1-Fluor-2,6-dichlorpyridiniumtetrafluoroborat 96%
1-Fluor-2,6-dichlorpyridiniumtetrafluoroborat

Electrophilic fluorinating agent.

Formel: C₅H₃BCl₂F₅N
Molecular Weight: 253,793 g/mol
Schmelzpunkt: 212…215 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00153157
CAS-Nummer: 140623-89-8
UN: 1759
ADR: 8,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR