2-Fluoroethyl methacrylate 97%

Lieferant: Apollo Scientific
Achtung

PC3751J-1G PC3751J-250MG
APOSPC3751J-1GEA 139 EUR
APOSPC3751J-1G APOSPC3751J-250MG
2-Fluoroethyl methacrylate 97%
2-Fluoroethyl methacrylate
Formel: C₆H₉FO₂
Molecular Weight: 132.14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00042288
CAS-Nummer: 686-54-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR