5-Fluorsalicylsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Fluor-2-hydroxybenzoesäure
PC3756F-25G
PC3756F-5G
PC3756F-100G
APOSPC3756F-25GEA
60.9
EUR
APOSPC3756F-25G
APOSPC3756F-5G
APOSPC3756F-100G
5-Fluorsalicylsäure 97%
5-Fluorsalicylsäure
Formel:
C₇H₅FO₃ Molecular Weight: 156,11 g/mol Schmelzpunkt: 177…182 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002456 CAS-Nummer: 345-16-4 EINECS: 206-455-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...