5-Fluoro-4-hydroxyquinazoline 97%

Lieferant: Apollo Scientific
Achtung

PC3756U-1G PC3756U-25G PC3756U-5G
APOSPC3756U-1GEA 21.9 EUR
APOSPC3756U-1G APOSPC3756U-25G APOSPC3756U-5G
5-Fluoro-4-hydroxyquinazoline 97%
5-Fluoroquinazolin-4(1H)-one
Formel: C₈H₅FN₂O
Molecular Weight: 164.14 g/mol
Schmelzpunkt: 238…242 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD20542997
CAS-Nummer: 436-72-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR