4-Amino-2,2-difluor-1,3-benzodioxol 98%

Lieferant: Apollo Scientific
Gefahr

PC3768-1G PC3768-5G
APOSPC3768-1GEA 72.5 EUR
APOSPC3768-1G APOSPC3768-5G
4-Amino-2,2-difluor-1,3-benzodioxol 98%
4-Amino-2,2-difluor-1,3-benzodioxol
Formel: C₇H₅F₂NO₂
Molecular Weight: 173,12 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00792416
CAS-Nummer: 106876-54-4
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR