3-Chloro-2-fluoroprop-1-ene 95%

Lieferant: Apollo Scientific
Achtung

PC3775-5G PC3775-1G PC3775-250MG
APOSPC3775-5GEA 621 EUR
APOSPC3775-5G APOSPC3775-1G APOSPC3775-250MG
3-Chloro-2-fluoroprop-1-ene 95%
3-Chloro-2-fluoroprop-1-ene
Formel: C₃H₄ClF
Molecular Weight: 94.52 g/mol
Siedepunkt: 55 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01696350
CAS-Nummer: 6186-91-0
UN: 1993
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR