2-Fluor-6-iodbenzonitril 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC3784-100G
PC3784-25G
PC3784-500G
PC3784-10G
PC3784-1KG
APOSPC3784-100GEA
126
EUR
APOSPC3784-100G
APOSPC3784-25G
APOSPC3784-500G
APOSPC3784-10G
APOSPC3784-1KG
2-Fluor-6-iodbenzonitril 98%
2-Fluor-6-iodbenzonitril
Formel:
C₇H₃FIN Molecular Weight: 247,01 g/mol Schmelzpunkt: 48…53 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015478 CAS-Nummer: 79544-29-9 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...