2-Fluor-5-iodbenzotrifluorid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Fluoro-4-iodo-2-(trifluoromethyl)benzene
PC3785E-25G
PC3785E-5G
APOSPC3785E-25GEA
78.3
EUR
APOSPC3785E-25G
APOSPC3785E-5G
2-Fluor-5-iodbenzotrifluorid
2-Fluor-5-iodbenzotrifluorid
Formel:
C₇H₃F₄I Molecular Weight: 290 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236678 CAS-Nummer: 59382-39-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...