3-Fluor-4-methoxybenzylalkohol 98%

Lieferant: Apollo Scientific
Achtung

PC3799-1G PC3799-5G
APOSPC3799-1GEA 33.5 EUR
APOSPC3799-1G APOSPC3799-5G
3-Fluor-4-methoxybenzylalkohol 98%
3-Fluor-4-methoxybenzylalkohol
Formel: C₈H₉FO₂
Molecular Weight: 156.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00070642
CAS-Nummer: 96047-32-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR