3H-Heptafluoro-2,2,4,4-tetrahydroxypentane 97%
Lieferant: Apollo Scientific
Synonyme:
1,1,1,3,5,5,5-Heptafluoroacetylacetone dihydrate
PC3806-1G
PC3806-5G
APOSPC3806-1GEA
357
EUR
APOSPC3806-1G
APOSPC3806-5G
3H-Heptafluoro-2,2,4,4-tetrahydroxypentane 97%
3H-Heptafluoro-2,2,4,4-tetrahydroxypentane
Formel:
C₅H₅F₇O₄ Molecular Weight: 262,08 g/mol Schmelzpunkt: 113…114 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02093308 CAS-Nummer: 77953-71-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...