2-Amino-4,6-difluorbenzothiazol

Lieferant: Apollo Scientific
Achtung

Synonyme: 4,6-Difluorobenzothiazol-2-ylamine

PC3812-5G PC3812-100G PC3812-25G
APOSPC3812-5GEA 42.2 EUR
APOSPC3812-5G APOSPC3812-100G APOSPC3812-25G
2-Amino-4,6-difluorbenzothiazol
2-Amino-4,6-difluorobenzothiazole
Formel: C₇H₄F₂N₂S
Molecular Weight: 186.18 g/mol
Schmelzpunkt: 216…220 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00276099
CAS-Nummer: 119256-40-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR