N-Morpholinium nonafluorobutanesulphonate
Lieferant: Apollo Scientific
PC3815-25G
PC3815-5G
APOSPC3815-25GEA
228
EUR
APOSPC3815-25G
APOSPC3815-5G
N-Morpholinium nonafluorobutanesulphonate
N-Morpholinium nonafluorobutanesulphonate
Formel:
C₈H₁₀F₉NO₄S Molecular Weight: 387,22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094228 CAS-Nummer: 503155-89-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...