Poly(chlortrifluorethylen) 800
Lieferant: Apollo Scientific
|
Achtung
|
PC3816A-1LB
PC3816A-4LB
APOSPC3816A-1LBEA
691
EUR
APOSPC3816A-1LB
APOSPC3816A-4LB
Poly(chlortrifluorethylen) 800
Poly(chlortrifluorethylen) 800
Formel:
(C₂ClF₃)n Molecular Weight: 800 g/mol Schmelzpunkt: >260 °C Lagertemperatur: Raumtemperatur |
CAS-Nummer:
9002-83-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...