2'-Fluor-5'-methylacetophenon 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2-Fluoro-5-methylphenyl)ethanone

PC3817KP-1G PC3817KP-5G PC3817KP-10G
APOSPC3817KP-1GEA 21.9 EUR
APOSPC3817KP-1G APOSPC3817KP-5G APOSPC3817KP-10G
2'-Fluor-5'-methylacetophenon 97%
2'-Fluor-5'-methylacetophenon
Formel: C₉H₉FO
Molecular Weight: 152.17 g/mol
Siedepunkt: 75…79 °C/0.7mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00236680
CAS-Nummer: 446-07-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR