2-Fluor-m-toluylsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Fluoro-3-methylbenzoic acid
PC3823-1G
PC3823-5G
APOSPC3823-1GEA
51
EUR
APOSPC3823-1G
APOSPC3823-5G
2-Fluor-m-toluylsäure 98%
2-Fluor-m-toluylsäure
Formel:
C₈H₇FO₂ Molecular Weight: 154.14 g/mol Schmelzpunkt: 114…116 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042504 CAS-Nummer: 315-31-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...