2-Fluoro-4-isocyanato-1-methylbenzene 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Fluoro-4-methylphenyl isocyanate

PC3823S-1G PC3823S-25G PC3823S-5G
APOSPC3823S-1GEA 31.9 EUR
APOSPC3823S-1G APOSPC3823S-25G APOSPC3823S-5G
2-Fluoro-4-isocyanato-1-methylbenzene 97%
2-Fluoro-4-isocyanato-1-methylbenzene

  • May form small amount of sediment.

Formel: C₈H₆FNO
Molecular Weight: 151,14 g/mol
Siedepunkt: 187…192 °C
Dichte: 1,175 g/cm³ (25 °C)
Flammpunkt: 79 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00037071
CAS-Nummer: 102561-42-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR