4-Fluor-1-naphthoesäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC3831N-1G
PC3831N-25G
PC3831N-5G
APOSPC3831N-1GEA
21.9
EUR
APOSPC3831N-1G
APOSPC3831N-25G
APOSPC3831N-5G
4-Fluor-1-naphthoesäure 97%
4-Fluor-1-naphthoesäure
Formel:
C₁₁H₇FO₂ Molecular Weight: 190,17 g/mol Schmelzpunkt: 224…227 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004011 CAS-Nummer: 573-03-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...