Perfluoroallyl iodide 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Perfluoro-3-iodoprop-1-ene

PC3834-1G PC3834-5G
APOSPC3834-1GEA 168 EUR
APOSPC3834-1G APOSPC3834-5G
Perfluoroallyl iodide 97%
Perfluoroallyl iodide

Useful for preparation of perfluoroallyl organometallics,Burton et al., J. Fluorine Chem., 1990,50,257

Formel: C₃F₅I
Molecular Weight: 257,93 g/mol
Siedepunkt: 53 °C
Dichte: 2,109 g/cm³ (25 °C)
Lagertemperatur: Kühlschrank
MDL: MFCD02093305
CAS-Nummer: 431-65-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR