Morpholinium hexafluorophosphate 97%

Lieferant: Apollo Scientific

PC3846-25G PC3846-5G
APOSPC3846-25GEA 245 EUR
APOSPC3846-25G APOSPC3846-5G
Morpholinium hexafluorophosphate 97%
Morpholinium hexafluorophosphate
Formel: C₄H₁₀F₆NOP
Molecular Weight: 233,09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00067328
CAS-Nummer: 25866-83-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR