2,2,3,3,3-Pentafluoropropylamine hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,2,3,3,3-Pentafluoropropan-1-amine hydrochloride

PC3851-1G PC3851-5G
APOSPC3851-1GEA 105 EUR
APOSPC3851-1G APOSPC3851-5G
2,2,3,3,3-Pentafluoropropylamine hydrochloride 97%
2,2,3,3,3-Pentafluoropropylamine hydrochloride
Formel: C₃H₅ClF₅N
Molecular Weight: 185,53 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00054747
CAS-Nummer: 374-14-1
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR